Möchten Sie sich wieder anmelden?
Panthera Biological Microscopes
NEW!
Panthera Biological Microscopes
Shift your lab to a new era
Panthera revolutionises the conventional microscope concept by creating a smart microscope. Its design, together with its new optics and digital capabilities, makes it the microscope of tomorrow.
  • New rackless and compact mechanical stage (patented) (not S model)
  • New plan UC objectives (not S and E models)
  • Motic Light Tracer: Coded nosepiece and digital light intensity knob (U-L-HD models)
  • Integrated USB for camera power
  • Sleep mode for lab safety and saving energy (U-L-HD) models
  • Smart Microscope ImagingOnDevice (for L & HD only)
View video
  Model Info Head Cat.No. Promo Price Quantity
  Panthera S S (school) Binocular 630-2936E  
  Panthera S S (school) Trinocular 630-2937E  
  Panthera E E (easyschool) Binocular 630-2940E  
  Panthera E E (easyschool) Trinocular 630-2941E  
  Panthera U U (university) Binocular 630-2932E  
  Panthera U U (university) Trinocular 630-2933E  
  Panthera C C (classic) Binocular 630-2938E  
  Panthera C C (classic) Trinocular 630-2939E  
  Panthera L L (lifescience) Binocular with integrated Smart
CA Mandimage On Device system
630-2931E  
  Panthera HD HD (highdefinition) Digitaluseonly:Integrated Smart
CA Mandimage On Device system
630-2935E