Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Möchten Sie sich wieder anmelden?
Chromatography columns, Omnifit®
OMNI006BCC-35-15AF OMNI006BCC-15-10AA OMNI006BCC-15-15FF OMNI006BCC-10-25AF OMNI006BCC-06-15FF OMNI006BCC-25-10FF OMNI006BCC-50-25AF OMNI006BCC-25-40FF OMNI006BCC-06-15AF OMNI006BCC-15-15AF OMNI006BCC-25-15FF OMNI006BCC-50-50AF OMNI006BCC-06-05FF OMNI006BCC-25-50FF OMNI006BCC-06-25FF OMNI006BCC-10-40AF OMNI006BCC-35-40AA OMNI006BCC-50-40FF OMNI006BCC-25-15AA OMNI006BCC-15-50FF OMNI006BCC-10-25AA OMNI006BCC-10-10AA OMNI006BCC-10-50AA OMNI006BCC-15-50AA OMNI006BCC-25-40AA OMNI006BCC-06-15AA OMNI006BCC-10-40FF OMNI006BCC-06-40AF OMNI006BCC-15-25AA OMNI006BCC-50-25AA OMNI006BCC-15-10FF OMNI006BCC-15-15AA OMNI006BCC-15-50AF OMNI006BCC-25-10AF OMNI006BCC-10-40AA OMNI006BCC-35-25AA OMNI006BCC-06-40FF OMNI006BCC-35-25AF OMNI006BCC-25-50AF OMNI006BCC-15-10AF OMNI006BCC-50-50AA OMNI006BCC-06-25AF OMNI006BCC-10-50FF OMNI006BCC-10-15FF OMNI006BCC-15-25AF OMNI006BCC-10-50AF OMNI006BCC-15-25FF OMNI006BCC-06-05AA OMNI006BCC-25-40AF OMNI006BCC-25-15AF OMNI006BCC-25-25AF OMNI006BCC-06-40AA OMNI006BCC-10-25FF OMNI006BCC-25-50AA OMNI006BCC-10-10AF OMNI006BCC-35-40AF OMNI006BCC-06-25AA OMNI006BCC-10-15AA OMNI006BCC-50-50FF OMNI006BCC-10-15AF OMNI006BCC-35-15FF OMNI006BCC-06-10AF OMNI006BCC-15-40FF OMNI006BCC-25-10AA OMNI006BCC-35-40FF OMNI006BCC-25-25FF OMNI006BCC-15-40AA OMNI006BCC-25-25AA OMNI006BCC-50-40AF OMNI006BCC-15-40AF OMNI006BCC-50-40AA OMNI006BCC-06-05AF
Chromatography columns, Omnifit®
These columns are designed to suit the majority of standard laboratory chromatography applications. They are ideal for aqueous systems and compatible with solvents used in common liquid chromatography applications such as protein purification.
  • Economical modular columns for low pressure or gravity feed, providing even distribution of eluent to the medium bed, with or without flow adapters
  • Borosilicate glass columns, PTFE endpieces
  • Compatible with a variety of bulk media
Operating temperature: 4 - 60 °C
Operating pressure: 6,6 mm 60 bar; 10 mm 40 bar; 15 mm 20 bar; 25, 35 and 50 mm 10 bar
pH stability: 1 - 14
Bestellen
BESTELLEN
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge