Bulb pipettes, PP
Lieferant: Vitlab
VITL164394EA 1670 EUR
VITL164394 VITL164494 VITL164194 VITL164294 VITL164594 VITL164094
Bulb pipettes, PP
Pipetten Vollpipetten
Highly transparent bulb pipettes made of PP.
  • Calibrated to deliver 'Ex'
  • With one high-contrast blue mark
  • Break-resistant
  • High chemical resistance
Please note: Exposure to temperatures above 60 °C can lead to volume changes. Recommended cleaning with mild alkaline detergents up to 60 °C.