Cap sealing mat, 384 Multisip septum cap mat
Lieferant: Thermo Scientific
ADVAAB-1171EA 119 EUR
ADVAAB-1171
Cap sealing mat, 384 Multisip septum cap mat
The 384 Multisip septum cap mat features a split septum design. This is perfect for sealing tall chimney 384-well PCR and storage plates. This mat can withstand a wide range of temperatures, and the product is 100% resealable, even after multiple piercings. Caps can be pierced simultaneously.
  • Suitable for PCR when used in conjunction with screw or clip-down heated lid thermal cyclers
  • Pierceable with standard pipetting equipment
  • 384 septum caps may be pierced simultaneously
  • Less than 0,5 N pierce pressure per cap
  • Solvent resistant, including acetonitrile and DMSO
Temperature range: –20 °C to +120 °C