Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')mangan(III), Sigma-Aldrich®
M-Clarity™ quality level = MQ100
SIAL700223-1GEA
84
EUR
SIAL700223-1G
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')mangan(III), Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')mangan(III)
Formel:
C₃₃H₅₇MnO₆ Molekulargewicht: 604,74 g/mol |
MDL Number:
MFCD00269825 CAS-Nummer: 14324-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...