2,1,3-Benzothiadiazol-4-ylamin
Lieferant: Thermo Scientific
MAYBKM06374.1EA
0
EUR
MAYBKM06374.1
2,1,3-Benzothiadiazol-4-ylamin
2,1,3-Benzothiadiazol-4-ylamin
Formel:
C₆H₅N₃S Molekulargewicht: 151,19 g/mol |
MDL Number:
MFCD00005810 CAS-Nummer: 767-64-6 EINECS: 212-186-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...