2-Amino-4-(4-bromphenyl)-5-methylthiazol, Sigma-Aldrich®
Lieferant: Merck
SIAL717851-1GEA
43.8
EUR
SIAL717851-1G
2-Amino-4-(4-bromphenyl)-5-methylthiazol, Sigma-Aldrich®
2-Amino-4-(4-bromophenyl)-5-methylthiazole
Formel:
C₁₀H₉BrN₂S Molekulargewicht: 269,16 g/mol |
MDL Number:
MFCD00170268 CAS-Nummer: 65705-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...