4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®
Lieferant: Sigma-Aldrich
M-Clarity™ quality level = MQ100
SAFA702528-1GEA
76.1
EUR
SAFA702528-1G
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Formel:
C₁₀H₈N₂O₂S Molekulargewicht: 220,25 |
MDL Number:
MFCD01316364 CAS-Nummer: 34418-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...