N-(4-Fluorobenzyl)-3,4-dimethylaniline, 97%

H57088.MDEA 61.3 EUR
ALFAH57088.MD ALFAH57088.03
N-(4-Fluorobenzyl)-3,4-dimethylaniline, 97%
Purity: 97%
CAS No.: 356531-47-0
Molecular Formula: C15H16FN
Formula Weight: 229.30
MDL No.: MFCD03210762
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR