3-Brom-6-fluorchromon, Sigma-Aldrich®
Lieferant: Sigma-Aldrich
M-Clarity™ quality level = MQ100
SAFA543675-5GEA
217
EUR
SAFA543675-5G
3-Brom-6-fluorchromon, Sigma-Aldrich®
3-Brom-6-fluorchromon
Formel:
C₉H₄BrFO₂ Molekulargewicht: 243,03 |
MDL Number:
MFCD03094003 CAS-Nummer: 179111-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...