3-Formyl-6-nitrochromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL402192-1GEA 32 EUR
SIAL402192-1G
3-Formyl-6-nitrochromon, Sigma-Aldrich®
3-Formyl-6-nitrochromon
Formel: C₁₀H₅NO₅
Molekulargewicht: 219,1504 g/mol
MDL Number: MFCD00192184
CAS-Nummer: 42059-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR