3-Formyl-6-methylchromon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL383473-5GEA
70.1
EUR
SIAL383473-5G
3-Formyl-6-methylchromon, Sigma-Aldrich®
3-Formyl-6-methylchromon
Formel:
C₁₁H₈O₃ Molekulargewicht: 188,1794 g/mol |
MDL Number:
MFCD00138943 CAS-Nummer: 42059-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...