Triethylphosphinsulfid
|
30392.04EA
158
EUR
30392.04
Triethylphosphinsulfid
Triethylphosphine sulphide
Formel:
C₆H₁₅PS |
MDL Number:
MFCD00014462 CAS-Nummer: 597-51-3 UN-Nummer: 3464 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...