Chlorbis(2-methoxyphenyl)phosphin, Sigma-Aldrich®

Lieferant: Sigma-Aldrich

SAFA710334-1GEA 161 EUR
SAFA710334-1G
Chlorbis(2-methoxyphenyl)phosphin, Sigma-Aldrich®
Chlorbis(2-methoxyphenyl)phosphin
Formel: C₁₄H₁₄ClO₂P
Molekulargewicht: 280,69
MDL Number: MFCD08064037
CAS-Nummer: 263369-88-6
UN-Nummer: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR