1H-Pyrazol-4-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL300713-500MGEA 126 EUR
SIAL300713-500MG
1H-Pyrazol-4-carbonsäure, Sigma-Aldrich®
1H-Pyrazole-4-carboxylic acid
Formel: C₄H₄N₂O₂
Molekulargewicht: 112,09
MDL Number: MFCD00011558
CAS-Nummer: 37718-11-9
EINECS: 253-641-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR