1-Phenyl-1H-pyrazol-5-carbonsäure
MAYBCC24501CBEA
0
EUR
MAYBCC24501CB
MAYBCC24501DA
1-Phenyl-1H-pyrazol-5-carbonsäure
1-Phenyl-1H-pyrazole-5-carboxylic acid
Formel:
C₁₀H₈N₂O₂ |
MDL Number:
MFCD00463580 CAS-Nummer: 1133-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...