1-Brom-4-(trimethylsilyl)benzol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL455113-5GEA
67.7
EUR
SIAL455113-5G
1-Brom-4-(trimethylsilyl)benzol, Sigma-Aldrich®
1-Brom-4-(trimethylsilyl)benzol
Formel:
C₉H₁₃BrSi Molekulargewicht: 229,1890 g/mol |
MDL Number:
MFCD00094165 CAS-Nummer: 6999-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...