2-Acetamido-5-chlorthiazol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL673129-10GEA
71
EUR
SIAL673129-10G
2-Acetamido-5-chlorthiazol, Sigma-Aldrich®
2-Acetamido-5-chlorthiazol
Formel:
C₅H₅ClN₂OS Molekulargewicht: 176,63 |
MDL Number:
MFCD01318098 CAS-Nummer: 20256-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...