Ethyl-2-amino-4-methylthiazol-5-carboxylat

Lieferant: MAYBRIDGE

MAYBRH01231EAEA 0 EUR
MAYBRH01231EA
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
Molekulargewicht: 186,22
MDL Number: MFCD00123414
CAS-Nummer: 7210-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR