Phenylglycidylether, Sigma-Aldrich®

Lieferant: Merck

SIAL578576-100GEA 68.8 EUR
SIAL578576-100G
Phenylglycidylether, Sigma-Aldrich®
Phenylglycidylether
Formel: C₉H₁₀O₂
Molekulargewicht: 150,18
MDL Number: MFCD00005133
CAS-Nummer: 122-60-1
EINECS: 204-557-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR