Ethylphenoxyacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL462934-25MLEA
42.1
EUR
SIAL462934-25ML
Ethylphenoxyacetat, Sigma-Aldrich®
Ethylphenoxyacetat
Formel:
C₁₀H₁₂O₃ Molekulargewicht: 180,20 g/mol |
MDL Number:
MFCD00026895 CAS-Nummer: 2555-49-9 EINECS: 219-867-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...