Thioacetanilid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL163651-10GEA 735 EUR
SIAL163651-10G
Thioacetanilid, Sigma-Aldrich®
Thioacetanilid
Formel: C₈H₉NS
MDL Number: MFCD00004942
CAS-Nummer: 637-53-6
EINECS: 211-288-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR