N-Ethyl-N-(3-sulfopropyl)-m-anisidin Natrium, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALE3013-500MGEA
253
EUR
SIALE3013-500MG
N-Ethyl-N-(3-sulfopropyl)-m-anisidin Natrium, Sigma-Aldrich®
N-Ethyl-N-(3-sulfopropyl)-m-anisidin Natrium
Formel:
C₁₂H₁₈NNaO₄S Molekulargewicht: 295,3304 g/mol |
MDL Number:
MFCD00083320 CAS-Nummer: 82611-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...