1-Methyl-5-phenyl-1H-pyrazol-4-carbaldehyd

MAYBCC28104DAEA 0 EUR
MAYBCC28104DA MAYBCC28104CB
1-Methyl-5-phenyl-1H-pyrazol-4-carbaldehyd
1-Methyl-5-phenyl-1H-pyrazol-4-carbaldehyd
Formel: C₁₁H₁₀N₂O
MDL Number: MFCD04115389
CAS-Nummer: 154927-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR