2'-Benzoyl-2,4'-dichloracetanilid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL561177-5GEA
147
EUR
SIAL561177-5G
2'-Benzoyl-2,4'-dichloracetanilid, Sigma-Aldrich®
2'-Benzoyl-2,4'-dichloracetanilid
Formel:
C₁₅H₁₁Cl₂NO₂ |
MDL Number:
MFCD00018905 CAS-Nummer: 4016-85-7 EINECS: 223-678-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...