3,5-Diformylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL567485-1GEA 504 EUR
SIAL567485-1G
3,5-Diformylphenylboronsäure, Sigma-Aldrich®
3,5-Diformylphenylboronsäure
Formel: C₈H₇BO₄
MDL Number: MFCD03701570
CAS-Nummer: 480424-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR