Potassium 3-methoxycarbonyl-5-nitrophenyltrifluoroborate, 96%

H52457.MDEA 181 EUR
ALFAH52457.MD ALFAH52457.03
Potassium 3-methoxycarbonyl-5-nitrophenyltrifluoroborate, 96%
Purity: 96%
CAS No.: 850623-56-2
Molecular Formula: C8H6BF3KNO4
Formula Weight: 287.04
Melting Point: 280-284°
MDL No.: MFCD04115749
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR