3-Brom-4-hydroxy-5-methoxybenzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL701165-5GEA
135
EUR
SIAL701165-5G
3-Brom-4-hydroxy-5-methoxybenzonitril, Sigma-Aldrich®
3-Brom-4-hydroxy-5-methoxybenzonitril
Formel:
C₈H₆BrNO₂ Molekulargewicht: 228,0400 g/mol |
MDL Number:
MFCD01123027 CAS-Nummer: 52805-45-5 UN-Nummer: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...