4-Cyanphenylhydrazin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL453471-5GEA
108
EUR
SIAL453471-5G
4-Cyanphenylhydrazin Hydrochlorid, Sigma-Aldrich®
4-Cyanphenylhydrazin Hydrochlorid
Formel:
C₇H₈ClN₃ Molekulargewicht: 169,61 g/mol |
MDL Number:
MFCD00673994 CAS-Nummer: 2863-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...