Methyl-3-methoxysalicylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL449490-10GEA 78.2 EUR
SIAL449490-10G
Methyl-3-methoxysalicylat, Sigma-Aldrich®
Methyl-3-methoxysalicylat
Formel: C₉H₁₀O₄
MDL Number: MFCD00239511
CAS-Nummer: 6342-70-7
EINECS: 228-737-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR