Dimethyl-4-methoxyphthalat, Sigma-Aldrich®

Lieferant: Merck

SIAL65250-10ML-FEA 360 EUR
SIAL65250-10ML-F
Dimethyl-4-methoxyphthalat, Sigma-Aldrich®
Dimethyl-4-methoxyphthalat
Formel: C₁₁H₁₂O₅
MDL Number: MFCD00077817
CAS-Nummer: 22895-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR