2-Methoxy-5-(trifluoromethoxy)benzonitrile, 97%

H26551.03EA 14.6 EUR
ALFAH26551.03 ALFAH26551.09
2-Methoxy-5-(trifluoromethoxy)benzonitrile, 97%
Purity: 97%
CAS No.: 886500-03-4
Molecular Formula: C9H6F3NO2
Formula Weight: 217.15
Refractive Index: 1.4710
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR