N-Cyclohexyl-2-fluorobenzamide, 97%

H56349.03EA 343 EUR
ALFAH56349.03 ALFAH56349.MD
N-Cyclohexyl-2-fluorobenzamide, 97%
Purity: 97%
CAS No.: 2267-95-0
Molecular Formula: C13H16FNO
Formula Weight: 221.27
MDL No.: MFCD00544069
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR