N-Cyclopentyl-2-fluorobenzamide, 97%

H56280.03EA 246 EUR
ALFAH56280.03 ALFAH56280.MD
N-Cyclopentyl-2-fluorobenzamide, 97%
Purity: 97%
CAS No.: 331435-48-4
Molecular Formula: C12H14FNO
Formula Weight: 207.25
MDL No.: MFCD00751457
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR