4-(Trifluormethoxy)phenylhydrazin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

M-Clarity™ quality level = MQ100

366951-1G
SIAL366951-1GEA 45.2 EUR
SIAL366951-1G
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid, Sigma-Aldrich®
4-(Trifluormethoxy)phenylhydrazin Hydrochlorid
Formel: C₇H₈ClF₃N₂O
Molekulargewicht: 228,60 g/mol
MDL Number: MFCD00053033
CAS-Nummer: 133115-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR