2-Amino-4-(4-chlorphenyl)thiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL339369-1GEA 28 EUR
SIAL339369-1G
2-Amino-4-(4-chlorphenyl)thiazol, Sigma-Aldrich®
2-Amino-4-(4-chlorophenyl)thiazole
Formel: C₉H₇ClN₂S
Molekulargewicht: 210,69
MDL Number: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR