2-Amino-4-(4-nitrophenyl)thiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL658499-10GEA 250 EUR
SIAL658499-10G
2-Amino-4-(4-nitrophenyl)thiazol, Sigma-Aldrich®
2-Amino-4-(4-nitrophenyl)thiazole
Formel: C₉H₇N₃O₂S
MDL Number: MFCD00170222
CAS-Nummer: 2104-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR