4-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-((Trimethylsilyl)ethynyl)benzaldehyde, 4-(Trimethylsilylethynyl)benzaldehyde
523380-5G
SIAL523380-5GEA
194
EUR
SIAL523380-5G
4-[(Trimethylsilyl)ethinyl]benzaldehyd, Sigma-Aldrich®
4-[(Trimethylsilyl)ethinyl]benzaldehyd
Formel:
C₁₂H₁₄OSi Molekulargewicht: 202,33 g/mol |
MDL Number:
MFCD02093765 CAS-Nummer: 77123-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...