4-Methyl-3-nitrophenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL521477-1GEA 62.4 EUR
SIAL521477-1G
4-Methyl-3-nitrophenylboronsäure, Sigma-Aldrich®
4-Methyl-3-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molekulargewicht: 180,96 g/mol
MDL Number: MFCD00191550
CAS-Nummer: 80500-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR