2-Bromo-N-(3-fluorobenzyl)-4-methylaniline, 97%
Lieferant: Thermo Scientific
H57207.MDEA 85.7 EUR
ALFAH57207.MD ALFAH57207.03
2-Bromo-N-(3-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1019587-76-8
Molecular Formula: C14H13BrFN
Formula Weight: 294.16