4-Bromo-N-(3-fluorobenzyl)-3-methylaniline, 97%

H57200.03EA 172 EUR
ALFAH57200.03 ALFAH57200.MD
4-Bromo-N-(3-fluorobenzyl)-3-methylaniline, 97%
Purity: 97%
CAS No.: 1019563-46-2
Molecular Formula: C14H13BrFN
Formula Weight: 294.16
MDL No.: MFCD11142809
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR