4-Bromo-N-(4-fluorobenzyl)-2-methylaniline, 97%

H57027.03EA 343 EUR
ALFAH57027.03 ALFAH57027.MD
4-Bromo-N-(4-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1019608-54-8
Molecular Formula: C14H13BrFN
Formula Weight: 294.16
MDL No.: MFCD11142043
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR