4-Chloro-N-(3-fluorobenzyl)-3-methylaniline, 96%

H57965.03EA 343 EUR
ALFAH57965.03 ALFAH57965.MD
4-Chloro-N-(3-fluorobenzyl)-3-methylaniline, 96%
Purity: 96%
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
MDL No.: MFCD22380731
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR