2-Fluoro-N-(3-fluorobenzyl)-3-methylaniline, 97%

H57346.03EA 191 EUR
ALFAH57346.03 ALFAH57346.MD
2-Fluoro-N-(3-fluorobenzyl)-3-methylaniline, 97%
Purity: 97%
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR