3-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%

H57957.MDEA 39 EUR
ALFAH57957.MD ALFAH57957.03
3-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1284974-45-3
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR