5-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%

H57171.MDEA 30.6 EUR
ALFAH57171.MD ALFAH57171.03
5-Fluoro-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1019568-49-0
Molecular Formula: C14H13F2N
Formula Weight: 233.26
MDL No.: MFCD11145912
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR