3-Fluoro-N-(4-fluorobenzyl)-4-methylaniline, 97%

H57784.MDEA 71.5 EUR
ALFAH57784.MD ALFAH57784.03
3-Fluoro-N-(4-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1019520-74-1
Molecular Formula: C14H13F2N
Formula Weight: 233.26
MDL No.: MFCD11146081
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR