Benzo[b]thiophen-5-carbaldehyd

Lieferant: Thermo Scientific

MAYBCC24804DEEA 0 EUR
MAYBCC24804DE MAYBCC24804DA
Benzo[b]thiophen-5-carbaldehyd
Benzo[b]thiophen-5-carbaldehyd
Formel: C₉H₆OS
Molekulargewicht: 162,2083 g/mol
MDL Number: MFCD05663673
CAS-Nummer: 10133-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR