3,5-Dichloro-N-(3-fluorobenzyl)aniline, 97%

H57062.03EA 172 EUR
ALFAH57062.03 ALFAH57062.MD
3,5-Dichloro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1040010-63-6
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11119642
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR