N-(3-Fluorobenzyl)-2-iodoaniline, 97%

H57098.MDEA 37.3 EUR
ALFAH57098.MD ALFAH57098.03
N-(3-Fluorobenzyl)-2-iodoaniline, 97%
Purity: 97%
CAS No.: 1039834-40-6
Molecular Formula: C13H11FIN
Formula Weight: 327.14
MDL No.: MFCD11179831
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR